MilliporeSigma
  • Determination of 255 pesticides in edible vegetable oils using QuEChERS method and gas chromatography tandem mass spectrometry.
Determination of 255 pesticides in edible vegetable oils using QuEChERS method and gas chromatography tandem mass spectrometry.
Analytical and bioanalytical chemistry (2016-11-14)
Zeying He, Yuehua Wang, Lu Wang, Yi Peng, Wenwen Wang, Xiaowei Liu
ABSTRACT
In this study, a simple and high-throughput method for determination of 255 pesticides in vegetable oils was developed based on QuEChERS sample preparation method combined with gas chromatography-triple quadrupole mass spectrometry. Different clean-up approaches were tested: A, 150 mg PSA + 150 mg C18; B, 250 mg PSA + 250 mg C18; C, 250 mg PSA + 250 mg C18 + 15 mg GCB; D, 250 mg PSA + 250 mg C18 + 50 mg GCB; and E, EMR-LipidTM. Best clean-up capacity was observed for EMR clean-up. The extraction procedures and parameters, including extraction time, solvent/sample ratio, and buffer system, were also thoroughly investigated and optimized. The limits of quantification (LOQ) ranged between 5 and 50 μg kg-1, and for the majority of the pesticides the LOQs were 5 μg kg-1, which were below the regulatory MRLs. Most recoveries at seven spiking levels were in the range of 70-120 % with RSDs <20 % indicating satisfactory accuracy. The coefficient of determination (r 2) was >0.99 within the calibration linearity range of 2-500 μg L-1 for the majority of the pesticides. This method was proved to be simple, sensitive, and effective, which can be applied for large-scale pesticide screening and quantification in vegetable oils.
Product Description
Supelco
Fenhexamid, PESTANAL®, analytical standard
Supelco
Fosthiazate, PESTANAL®, analytical standard
Supelco
Prothiofos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland