MilliporeSigma
  • Epidermal growth factor signaling protects from cholestatic liver injury and fibrosis.
Epidermal growth factor signaling protects from cholestatic liver injury and fibrosis.
Journal of molecular medicine (Berlin, Germany) (2016-08-29)
Jasmin Svinka, Sandra Pflügler, Markus Mair, Hanns-Ulrich Marschall, Jan G Hengstler, Patricia Stiedl, Valeria Poli, Emilio Casanova, Gerald Timelthaler, Maria Sibilia, Robert Eferl
ABSTRACT
We have demonstrated that the signal transducer and activator of transcription 3 (STAT3) protects from cholestatic liver injury. Specific ablation of STAT3 in hepatocytes and cholangiocytes (STAT3∆hc) aggravated liver damage and fibrosis in the Mdr2-/- (multidrug resistance 2) mouse model for cholestatic disease. Upregulation of bile acid biosynthesis genes and downregulation of epidermal growth factor receptor (EGFR) expression were observed in STAT3∆hc Mdr2-/- mice but the functional consequences of these processes in cholestatic liver injury remained unclear. Here, we show normal canalicular architecture and bile flow but increased amounts of bile acids in the bile of STAT3∆hc Mdr2-/- mice. Moreover, STAT3-deficient hepatocytes displayed increased sensitivity to bile acid-induced apoptosis in vitro. Since EGFR signaling has been reported to protect hepatocytes from bile acid-induced apoptosis, we generated mice with hepatocyte/cholangiocyte-specific ablation of EGFR (EGFR∆hc) and crossed them to Mdr2-/- mice. Importantly, deletion of EGFR phenocopied deletion of STAT3 and led to aggravated liver damage, liver fibrosis, and hyperproliferation of K19+ cholangiocytes. Our data demonstrate hepatoprotective functions of the STAT3-EGFR signaling axis in cholestatic liver disease. STAT3 is a negative regulator of bile acid biosynthesis. STAT3 protects from bile acid-induced apoptosis and regulates EGFR expression. EGFR signaling protects from cholestatic liver injury and fibrosis.
Sigma-Aldrich
Insulin-like Growth Factor-II human, IGF-II, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, ≥90% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.