MilliporeSigma
Dopaminergic dynamics underlying sex-specific cocaine reward.
Nature communications (2017-01-11)
Erin S Calipari, Barbara Juarez, Carole Morel, Deena M Walker, Michael E Cahill, Efrain Ribeiro, Ciorana Roman-Ortiz, Charu Ramakrishnan, Karl Deisseroth, Ming-Hu Han, Eric J Nestler
ABSTRACT
Although both males and females become addicted to cocaine, females transition to addiction faster and experience greater difficulties remaining abstinent. We demonstrate an oestrous cycle-dependent mechanism controlling increased cocaine reward in females. During oestrus, ventral tegmental area (VTA) dopamine neuron activity is enhanced and drives post translational modifications at the dopamine transporter (DAT) to increase the ability of cocaine to inhibit its function, an effect mediated by estradiol. Female mice conditioned to associate cocaine with contextual cues during oestrus have enhanced mesolimbic responses to these cues in the absence of drug. Using chemogenetic approaches, we increase VTA activity to mechanistically link oestrous cycle-dependent enhancement of VTA firing to enhanced cocaine affinity at DAT and subsequent reward processing. These data have implications for sexual dimorphism in addiction vulnerability and define a mechanism by which cellular activity results in protein alterations that contribute to dysfunctional learning and reward processing.
Sigma-Aldrich
Anti-Dopamine Transporter; 100 µg Antibody, from rabbit