MilliporeSigma
  • Adiabatic electron affinity of pentacene and perfluoropentacene molecules studied by anion photoelectron spectroscopy: Molecular insights into electronic properties.
Adiabatic electron affinity of pentacene and perfluoropentacene molecules studied by anion photoelectron spectroscopy: Molecular insights into electronic properties.
The Journal of chemical physics (2017-01-05)
Tsugunosuke Masubuchi, Yoshito Sugawara, Atsushi Nakajima
ABSTRACT
Pentacene (C
Sigma-Aldrich
6,13-Pentacenequinone, 99%