MilliporeSigma
  • Binding of N-methylscopolamine to the extracellular domain of muscarinic acetylcholine receptors.
Binding of N-methylscopolamine to the extracellular domain of muscarinic acetylcholine receptors.
Scientific reports (2017-01-17)
Jan Jakubík, Alena Randáková, Pavel Zimčík, Esam E El-Fakahany, Vladimír Doležal
ABSTRACT
Interaction of orthosteric ligands with extracellular domain was described at several aminergic G protein-coupled receptors, including muscarinic acetylcholine receptors. The orthosteric antagonists quinuclidinyl benzilate (QNB) and N-methylscopolamine (NMS) bind to the binding pocket of the muscarinic acetylcholine receptor formed by transmembrane α-helices. We show that high concentrations of either QNB or NMS slow down dissociation of their radiolabeled species from all five subtypes of muscarinic acetylcholine receptors, suggesting allosteric binding. The affinity of NMS at the allosteric site is in the micromolar range for all receptor subtypes. Using molecular modelling of the M