MilliporeSigma
  • Characterization of the human gene encoding the type I alpha and type I beta cGMP-dependent protein kinase (PRKG1).
Characterization of the human gene encoding the type I alpha and type I beta cGMP-dependent protein kinase (PRKG1).
Genomics (1997-06-01)
S Orstavik, V Natarajan, K Taskén, T Jahnsen, M Sandberg
ABSTRACT
The type I cGMP-dependent protein kinase (cGK) has been shown to play a crucial role in the relaxation of vascular smooth muscle by lowering the intracellular level of calcium. Two isoforms of type I cGK have been described, type I alpha and type I beta, differing only in their N-terminal parts. This report describes the cloning of the gene PRKG1 encoding both human type I cGK isoforms. PRKG1 is a single-copy gene consisting of 19 exons encompassing at least 220 kb. Several of the splice sites previously observed in the Drosophila melanogaster DG2 gene have been conserved in PRKG1, and these conserved splice sites correlated well with the boundaries between several of the previously proposed functional domains of type I cGK. The first two exons of the type I cGK gene were shown to encode the type I alpha- and type I beta-specific parts of the cGK. Using 5'-rapid amplification of cDNA ends, potential sites for transcription initiation were identified 5' upstream of both these exons. Northern blot analyses demonstrated distinct patterns of expression of the isoforms of type I alpha and I beta cGK in different human tissues.
Sigma-Aldrich
PRKG1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution