MilliporeSigma
  • Long-term stability improvement of light-emitting diode using highly transparent graphene oxide paste.
Long-term stability improvement of light-emitting diode using highly transparent graphene oxide paste.
Nanoscale (2016-10-08)
Seungae Lee, Yun Ki Kim, Jyongsik Jang
ABSTRACT
A highly transparent paste adhesive is successfully fabricated by introducing graphene oxide (GO) to silicone paste adhesive by using a solvent-exchange method. The GO incorporated in the paste adhesive has a significant role in improving thermal conductivity, transparency and adhesive strength. The GO-embedded silicone paste is applied as a die-attach paste to light-emitting diodes (LEDs) in order to enhance the optical quality of the LEDs. The presence of GO in the die-attach layer of the LEDs gives rise to the enhancement of luminous intensity, effective heat dissipation, improvement of moisture barrier property as well as high adhesive strength. Consequently, the LEDs with the GO-embedded die-attach paste exhibit enhanced long-term stability. This novel approach provides a feasible and effective strategy for improving LED performance.
Sigma-Aldrich
Graphene oxide, paste, non-exfoliated