MilliporeSigma
  • Induction of pluripotent stem cells by defined factors is greatly improved by small-molecule compounds.
Induction of pluripotent stem cells by defined factors is greatly improved by small-molecule compounds.
Nature biotechnology (2008-06-24)
Danwei Huangfu, René Maehr, Wenjun Guo, Astrid Eijkelenboom, Melinda Snitow, Alice E Chen, Douglas A Melton
ABSTRACT
Reprogramming of mouse and human somatic cells can be achieved by ectopic expression of transcription factors, but with low efficiencies. We report that DNA methyltransferase and histone deacetylase (HDAC) inhibitors improve reprogramming efficiency. In particular, valproic acid (VPA), an HDAC inhibitor, improves reprogramming efficiency by more than 100-fold, using Oct4-GFP as a reporter. VPA also enables efficient induction of pluripotent stem cells without introduction of the oncogene c-Myc.
Sigma-Aldrich
BIX 01294 trihydrochloride hydrate, ≥98% (HPLC), powder