MilliporeSigma
  • Anticancer drugs from nature--natural products as a unique source of new microtubule-stabilizing agents.
Anticancer drugs from nature--natural products as a unique source of new microtubule-stabilizing agents.
Natural product reports (2007-03-29)
Karl-Heinz Altmann, Jürg Gertsch
ABSTRACT
This review article provides an overview on the current state of research in the area of microtubule-stabilizing agents from natural sources, with a primary focus on the biochemistry, biology, and pharmacology associated with these compounds. A variety of natural products have been discovered over the last decade to inhibit human cancer cell proliferation through a taxol-like mechanism. These compounds represent a whole new range of structurally diverse lead structures for anticancer drug discovery.
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder