MilliporeSigma
  • Adsorptive voltammetry of 2-(5-bromo-2-pyridyl)azo-5-[N-n-propyl-N-(3-sulfopropyl)amino]phenol on a carbon paste electrode in the presence of organic cations and polycation.
Adsorptive voltammetry of 2-(5-bromo-2-pyridyl)azo-5-[N-n-propyl-N-(3-sulfopropyl)amino]phenol on a carbon paste electrode in the presence of organic cations and polycation.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-12-13)
Toshiaki Hattori, Naoko Tsurumi, Ryo Kato, Masanori Nakayama
ABSTRACT
The adsorption of ion-association complexes on a carbon paste electrode (CPE) was investigated by cyclic voltammetry using an electroactive hydrophobic anion probe. The redox reactions of 2-(5-bromo-2-pyridyl)azo-5-[N-n-propyl-N-(3-sulfopropyl)amino]phenol (5-Br-PAPS), the analytical probe, were irreversible. The reduction of the azo group and the oxidation of the phenol were observed at -0.1 V and 0.9 V vs. SCE, respectively, in a 0.1 mol L(-1) H(2)SO(4) solution. The peak currents for the redox reaction increased with the concentration of the cationic surfactant and the accumulation time. The increase in the ratio of the peak current to the concentration of cationic surfactants was proportional to the hydrophobicity. The peak current for 5-Br-PAPS also increased when a polycation, polyhexamethylene biguanide hydrochloride, was added and was strongly dependent on the ionic strength and pH, in contrast to cationic surfactants.
Product Description
Supelco
2-(5-Bromo-2-pyridylazo)-5-[N-propyl-N-(3-sulfopropyl)amino]phenol disodium salt dihydrate, for spectrophotometric det. of Zn(II), Cu(II), Fe(II), Co(II), H2O2, ≥95.0% (HPLC)