MilliporeSigma
  • The IASLC Mesothelioma Staging Project: Proposals for the M Descriptors and for Revision of the TNM Stage Groupings in the Forthcoming (Eighth) Edition of the TNM Classification for Mesothelioma.
The IASLC Mesothelioma Staging Project: Proposals for the M Descriptors and for Revision of the TNM Stage Groupings in the Forthcoming (Eighth) Edition of the TNM Classification for Mesothelioma.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer (2016-10-01)
Valerie W Rusch, Kari Chansky, Hedy L Kindler, Anna K Nowak, Harvey I Pass, David C Rice, Lynn Shemanski, Françoise Galateau-Sallé, Brian C McCaughan, Takashi Nakano, Enrico Ruffini, Jan P van Meerbeeck, Masahiro Yoshimura
ABSTRACT
The M component and TNM stage groupings for malignant pleural mesothelioma (MPM) have been empirical. The International Association for the Study of Lung Cancer developed a multinational database to propose evidence-based revisions for the eighth edition of the TNM classification of MPM. Data from 29 centers were submitted either electronically or by transfer of existing institutional databases. The M component as it currently stands was validated by confirming sufficient discrimination (by Kaplan-Meier analysis) with respect to overall survival (OS) between the clinical M0 (cM0) and cM1 categories. Candidate stage groups were developed by using a recursive partitioning and amalgamation algorithm applied to all cM0 cases. Of 3519 submitted cases, 2414 were analyzable and 84 were cM1 cases. Median OS for cM1 cases was 9.7 months versus 13.4 months (p = 0.0013) for the locally advanced (T4 or N3) cM0 cases, supporting inclusion of only cM1 in the stage IV group. Exploratory analyses suggest a possible difference in OS for single- versus multiple-site cM1 cases. A recursive partitioning and amalgamation-generated survival tree on the OS outcomes restricted to cM0 cases with the newly proposed (eighth edition) T and N components indicates that optimal stage groupings for the eighth edition will be as follows: stage IA (T1N0), stage IB (T2-3N0), stage II (T1-2N1), stage IIIA (T3N1), stage IIIB (T1-3N2 or any T4), and stage IV (any M1). This first evidence-based revision of the TNM classification for MPM leads to substantial changes in the T and N components and the stage groupings.