MilliporeSigma
  • Anti-hepatocellular carcinoma properties of the anti-alcoholism drug disulfiram discovered to enzymatically inhibit the AMPK-related kinase SNARK in vitro.
Anti-hepatocellular carcinoma properties of the anti-alcoholism drug disulfiram discovered to enzymatically inhibit the AMPK-related kinase SNARK in vitro.
Oncotarget (2016-09-08)
Kaku Goto, Naoya Kato, Raymond T Chung
ABSTRACT
We recently described that the anti-apoptotic AMPK-related kinase, SNARK, promotes transforming growth factor (TGF)-β signaling in hepatocellular carcinoma (HCC) cells, as a potentially new therapeutic target. Here we explored FDA-approved drugs inhibiting the enzymatic activity of SNARK, using an in vitro luminescence kinase assay system. Interestingly, the long-used anti-alcoholism drug disulfiram (DSF), also known as Antabuse, emerged as the top hit. Enzymatic kinetics analyses revealed that DSF inhibited SNARK kinase activity in a noncompetitive manner to ATP or phosphosubstrates. Comparative in vitro analyses of DSF analogs indicated the significance of the disulfide bond-based molecular integrity for the kinase inhibition. DSF suppressed SNARK-promoted TGF-β signaling and demonstrated anti-HCC effects. The chemical and enzymatic findings herein reveal novel pharmacological effects of and use for DSF and its derivatives, and could be conducive to prevention and inhibition of liver fibrosis and HCC.
Sigma-Aldrich
cis-11-Methyl-2-dodecenoic acid, ≥90.0% (HPLC)