MilliporeSigma
  • Differential protein expression in metallothionein protection from depleted uranium-induced nephrotoxicity.
Differential protein expression in metallothionein protection from depleted uranium-induced nephrotoxicity.
Scientific reports (2016-12-15)
Yuhui Hao, Jiawei Huang, Cong Liu, Hong Li, Jing Liu, Yiping Zeng, Zhangyou Yang, Rong Li
ABSTRACT
The purpose of this study was to investigate the underlying mechanism of metallothionein (MT) protection from depleted uranium (DU) using a proteomics approach to search for a DU toxicity-differential protein. MT-/- and MT+/+ mice were administrated with a single dose of DU (10 mg/kg, i.p.) or equal volume of saline. After 4 days, protein changes in kidney tissues were evaluated using a proteomics approach. A total of 13 differentially expressed proteins were identified using two-dimensional electrophoresis and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry. The validating results showed that the expression of aminoacylase-3 (ACY-3) and the mitochondrial ethylmalonic encephalopathy 1 (ETHE1) decreased significantly after DU exposure; in addition, the reduction in MT-/- mice was more significant than that in MT+/+ mice. The results also showed that exogenous ETHE1 or ACY-3 could increase the survival rate of human embryonic kidney 293 (HEK293) cells after DU exposure. A specific siRNA of ETHE1 significantly increased cell apoptosis rates after DU exposure, whereas exogenous ETHE1 significantly decreased cell apoptosis rates. In summary, ACY-3 and ETHE1 might involve in protection roles of MT. ETHE1 could be a new sensitive molecular target of DU-induced cell apoptosis.