MilliporeSigma
  • Puerarin Prevents LPS-Induced Osteoclast Formation and Bone Loss via Inhibition of Akt Activation.
Puerarin Prevents LPS-Induced Osteoclast Formation and Bone Loss via Inhibition of Akt Activation.
Biological & pharmaceutical bulletin (2016-12-03)
Yun Zhang, Ming Yan, Qing-Feng Yu, Pei-Fan Yang, Hai-Dong Zhang, Yong-Hong Sun, Zhi-Fen Zhang, Yun-Feng Gao
ABSTRACT
Osteolysis induced by chronic Gram-negative bacterial infection underlies many bone diseases such as osteomyelitis, septic arthritis, and periodontitis. Drugs that inhibit lipopolysaccharide (LPS)-induced osteolysis are critically needed for the prevention of bone destruction in infective bone diseases. In this study, we assessed the effect of puerarin, a natural isoflavone isolated from Pueraria lobata OHWI root, on LPS-induced osteoclastogenesis and bone loss. Our in vitro study showed that puerarin significantly inhibited LPS-induced osteoclast differentiation from osteoclast precursor RAW264.7 cells. The inhibition occurred through suppressing the production of osteoclast activating factor tumor necrosis factor (TNF)-α, interleukin (IL)-1β and prostaglandin E2 (PGE2), which led to down-regulating mRNA expression of osteoclastogenic genes including tartrate-resistant acid phosphatase (TRAP), cathepsin K and matrix metalloprotein 9 (MMP-9). Furthermore, LPS triggered activation of Akt in osteoclast precursor RAW264.7 cells, which was inhibited by puerarin treatment. In vivo, puerarin attenuated LPS-induced bone loss in a murine calvarial osteolysis model. Collectively, puerarin prevents LPS-induced osteoclast formation, function and bone loss, where the inhibition of Akt activation plays an important role. These findings provide evidences that puerarin might be beneficial as a promising candidate drug for the prevention and treatment of bacteria-induced bone destruction disease, and give new insights for understanding its possible mechanism.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Leukocyte Acid Phosphatase (TRAP) Kit, Kit formulated with all liquid reagents
Pricing and availability is not currently available.