MilliporeSigma
  • Residue analysis of thyreostats in baby foods via matrix solid phase dispersion and liquid chromatography - dual-polarity electrospray - tandem mass spectrometry.
Residue analysis of thyreostats in baby foods via matrix solid phase dispersion and liquid chromatography - dual-polarity electrospray - tandem mass spectrometry.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2016-10-04)
Alessandra Gentili, Valeria De Pratti, Fulvia Caretti, Virginia Pérez-Fernández, Pierpaolo Tomai, Roberta Curini, Lucia Mainero Rocca
ABSTRACT
This paper describes a rapid method for confirming residues of thyreostats in meat-based baby foods by using liquid chromatography - dual polarity electrospray - tandem mass spectrometry (LC-ES(±)-MS/MS). Six thioureylenes, belonging to the group of thiouracil and imidazole, were selected for this work: thiouracil (TU), methylthiouracil (MTU), propylthiouracil (PTU), phenylthiouracil (PhTU), mercaptobenzimidazole (MBI) and tapazole (TAP). The amphoteric nature of these compounds allows their electrospray detection in both positive and negative ionisation. Nevertheless, MS detection is not favoured by their low molecular weights, while their chromatographic retention is also thwarted by their high polarity. A pentafluorophenyl (PFP) core-shell phase column was selected to avoid peak asymmetry or peak splitting, and a dual-polarity ionisation method was optimised to obtain a sensitivity as high as possible. The method was validated according to the Commission Decision 657/2002/EC. A simple and fast procedure based on matrix solid phase dispersion (MSPD) was optimised to extract analytes from baby foods with recoveries exceeding 82%. Limit of decision (CCα) and detection capability (CCβ) were lower than the permissible maximum concentration (10 ng g-1). The validated method was then applied to assess the potential occurrence of the six selected thyreostats in nine commercial products. All the samples were found free of contamination.
Sigma-Aldrich
5-Propyl-2-thiouracil, ≥98%