MilliporeSigma
  • Highly selective inhibition of myosin motors provides the basis of potential therapeutic application.
Highly selective inhibition of myosin motors provides the basis of potential therapeutic application.
Proceedings of the National Academy of Sciences of the United States of America (2016-11-07)
Serena Sirigu, James J Hartman, Vicente José Planelles-Herrero, Virginie Ropars, Sheila Clancy, Xi Wang, Grace Chuang, Xiangping Qian, Pu-Ping Lu, Edward Barrett, Karin Rudolph, Christopher Royer, Bradley P Morgan, Enrico A Stura, Fady I Malik, Anne M Houdusse
ABSTRACT
Direct inhibition of smooth muscle myosin (SMM) is a potential means to treat hypercontractile smooth muscle diseases. The selective inhibitor CK-2018571 prevents strong binding to actin and promotes muscle relaxation in vitro and in vivo. The crystal structure of the SMM/drug complex reveals that CK-2018571 binds to a novel allosteric pocket that opens up during the "recovery stroke" transition necessary to reprime the motor. Trapped in an intermediate of this fast transition, SMM is inhibited with high selectivity compared with skeletal muscle myosin (IC50 = 9 nM and 11,300 nM, respectively), although all of the binding site residues are identical in these motors. This structure provides a starting point from which to design highly specific myosin modulators to treat several human diseases. It further illustrates the potential of targeting transition intermediates of molecular machines to develop exquisitely selective pharmacological agents.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder