MilliporeSigma
  • Molecular and cellular effects of vitamin B12 forms on human trophoblast cells in presence of excessive folate.
Molecular and cellular effects of vitamin B12 forms on human trophoblast cells in presence of excessive folate.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-10-04)
Tejas Shah, Kalpana Joshi, Sanjay Mishra, Suhas Otiv, Vijay Kumbar
ABSTRACT
Folic acid (FA) and iron are essential supplements during pregnancy. Similarly effects of vitamin B12 (B12) inadequacy and high folate and low B12 status, on pregnancy outcome are available. However there are no mandatory recommendations for B12. There are many forms of B12 viz. Cyanocobalamin (Cbl), Methylcobalamin (MeCbl), Adenosylcobalamin (AdCbl), and Hydroxycobalamin (HCbl) though there is limited consensus on which form has better efficacy. In the present study we have determined effect of various forms of B12 in the presence of two FA concentrations namely normal physiological (20ng/mL; NPFA) and supra-physiological (2000ng/mL; SPFA) concentration to mimic real time situation where FA is in excess due to supplementation. We assessed trophoblastic proliferation, viability, TNFα and EGFr mRNA expression, homocysteine, β-hCG and MDA levels. Trophoblastic viability was significantly suppressed at SPFA concentration and was restored by B12 treatment with Cbl, AdCbl and combination of MeCbl+AdCbl. The mRNA expressions of TNFα were up-regulated, while EGFr were down-regulated at SPFA concentrations, as validated by RT-PCR. Treatment with MeCbl+AdCbl significantly decreased homocysteine and MDA levels at SPFA concentrations. High levels of FA alone had a detrimental effect on placental health and functions as reflected by decreased viability, EGFr expression and increased TNFα expression, homocysteine and MDA levels. Combination of B12 active forms i.e. MeCbl+AdCbl was found to be most effective in neutralising excess folate effect in-vitro.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, 10 ×, With 1000 mg/L glucose (1x), without L-glutamine, sodium bicarbonate, and folic acid, liquid, sterile-filtered, suitable for cell culture