MilliporeSigma
  • Normobaric hyperoxia stimulates superoxide and nitric oxide production in the caudal solitary complex of rat brain slices.
Normobaric hyperoxia stimulates superoxide and nitric oxide production in the caudal solitary complex of rat brain slices.
American journal of physiology. Cell physiology (2016-10-14)
Geoffrey E Ciarlone, Jay B Dean
ABSTRACT
Central CO2-chemosensitive neurons in the caudal solitary complex (cSC) are stimulated not only by hypercapnic acidosis, but by hyperoxia as well. While a cellular mechanism for the CO2 response has yet to be isolated, previous data show that a redox-sensitive mechanism underlies neuronal excitability to hyperoxia. However, it remains unknown how changes in Po2 affect the production of reactive oxygen and nitrogen species (RONS) in the cSC that can lead to increased cellular excitability and, with larger doses, to cellular dysfunction and death. To this end, we used fluorescence microscopy in real time to determine how normobaric hyperoxia increases the production of key RONS in the cSC. Because neurons in the region are CO2 sensitive, we also examined the potential effects of CO2 narcosis, used during euthanasia before brain slice harvesting, on RONS production. Our findings show that normobaric hyperoxia (0.4 → 0.95 atmospheres absolute O2) increases the fluorescence rates of fluorogenic dyes specific to both superoxide and nitric oxide. Interestingly, different results were seen for superoxide fluorescence when CO2 narcosis was used during euthanasia, suggesting long-lasting changes in superoxide production and/or antioxidant activity subsequent to CO2 narcosis before brain slicing. Further research needs to distinguish whether the increased levels of RONS reported here are merely increases in oxidative and nitrosative signaling or, alternatively, evidence of redox and nitrosative stress.
Sigma-Aldrich
7-Nitroindazole, ≥98%