MilliporeSigma
  • Genetic architecture of nonadditive inheritance in Arabidopsis thaliana hybrids.
Genetic architecture of nonadditive inheritance in Arabidopsis thaliana hybrids.
Proceedings of the National Academy of Sciences of the United States of America (2016-11-03)
Danelle K Seymour, Eunyoung Chae, Dominik G Grimm, Carmen Martín Pizarro, Anette Habring-Müller, François Vasseur, Barbara Rakitsch, Karsten M Borgwardt, Daniel Koenig, Detlef Weigel
ABSTRACT
The ubiquity of nonparental hybrid phenotypes, such as hybrid vigor and hybrid inferiority, has interested biologists for over a century and is of considerable agricultural importance. Although examples of both phenomena have been subject to intense investigation, no general model for the molecular basis of nonadditive genetic variance has emerged, and prediction of hybrid phenotypes from parental information continues to be a challenge. Here we explore the genetics of hybrid phenotype in 435 Arabidopsis thaliana individuals derived from intercrosses of 30 parents in a half diallel mating scheme. We find that nonadditive genetic effects are a major component of genetic variation in this population and that the genetic basis of hybrid phenotype can be mapped using genome-wide association (GWA) techniques. Significant loci together can explain as much as 20% of phenotypic variation in the surveyed population and include examples that have both classical dominant and overdominant effects. One candidate region inherited dominantly in the half diallel contains the gene for the MADS-box transcription factor AGAMOUS-LIKE 50 (AGL50), which we show directly to alter flowering time in the predicted manner. Our study not only illustrates the promise of GWA approaches to dissect the genetic architecture underpinning hybrid performance but also demonstrates the contribution of classical dominance to genetic variance.
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)