MilliporeSigma
  • Redox-Neutral Aromatization of Cyclic Amines: Mechanistic Insights and Harnessing of Reactive Intermediates for Amine α- and β-C-H Functionalization.
Redox-Neutral Aromatization of Cyclic Amines: Mechanistic Insights and Harnessing of Reactive Intermediates for Amine α- and β-C-H Functionalization.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-10-07)
Longle Ma, Anirudra Paul, Martin Breugst, Daniel Seidel
ABSTRACT
Cyclic amines such as pyrrolidine and piperidine are known to undergo condensations with aldehydes to furnish pyrrole and pyridine derivatives, respectively. A combined experimental and computational study provides detailed insights into the mechanism of pyrrole formation. A number of reactive intermediates (e.g., azomethine ylides, conjugated azomethine ylides, enamines) were intercepted, outlining strategies for circumventing aromatization as a valuable pathway for amine C-H functionalization.
Sigma-Aldrich
Hexamethyleneimine, 99%