MilliporeSigma
  • Characterization, classification and authentication of fruit-based extracts by means of HPLC-UV chromatographic fingerprints, polyphenolic profiles and chemometric methods.
Characterization, classification and authentication of fruit-based extracts by means of HPLC-UV chromatographic fingerprints, polyphenolic profiles and chemometric methods.
Food chemistry (2016-12-17)
Naiara Pardo-Mates, Alex Vera, Sergio Barbosa, Míriam Hidalgo-Serrano, Oscar Núñez, Javier Saurina, Santiago Hernández-Cassou, Lluis Puignou
ABSTRACT
HPLC-UV was applied to the analysis and characterization of fruit-based and fruit-processed products. A Kinetex C18 reversed-phase column was proposed under gradient elution for the determination of 17 polyphenols. Acceptable sensitivity (LODs below 0.16mg/L), and good linearity (r2 higher than 0.995), precision (RSD below 6.8%), and method trueness (relative errors below 11%) were obtained. Data corresponding to polyphenolic peak areas and HPLC-UV chromatographic fingerprints were then analyzed by exploratory principal component analysis (PCA) to extract information of the most significant variables contributing to characterization and classification of analyzed samples regarding the fruit of origin. HPLC-UV chromatographic data was further treated by partial least square (PLS) regression to determine the percentages of adulteration in cranberry-fruit extracts. It was found that even mixture samples containing low percentages of adulterants could be distinguished from genuine cranberry extracts. Highly satisfactory results were obtained, with overall errors in the quantification of adulterations below 4.3%.
Sigma-Aldrich
Quercitrin hydrate, ≥78%
Sigma-Aldrich
Ethyl gallate, antioxidant, ≥96.0% (HPLC)