MilliporeSigma
Lens extrusion from Laminin alpha 1 mutant zebrafish.
TheScientificWorldJournal (2014-02-15)
Mallika Pathania, Elena V Semina, Melinda K Duncan
ABSTRACT
We report analysis of the ocular lens phenotype of the recessive, larval lethal zebrafish mutant, lama1 (a69/a69). Previous work revealed that this mutant has a shortened body axis and eye defects including a defective hyaloid vasculature, focal corneal dysplasia, and loss of the crystalline lens. While these studies highlight the importance of laminin α1 in lens development, a detailed analysis of the lens defects seen in these mutants was not reported. In the present study, we analyze the lenticular anomalies seen in the lama1 (a69/a69) mutants and show that the lens defects result from the anterior extrusion of lens material from the eye secondary to structural defects in the lens capsule and developing corneal epithelium associated with basement membrane loss. Our analysis provides further insights into the role of the lens capsule and corneal basement membrane in the structural integrity of the developing eye.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Aquaporin 0 Antibody, Chemicon®, from rabbit