MilliporeSigma
  • Deficits in receptor-mediated endocytosis and recycling in cells from mice with Gpr107 locus disruption.
Deficits in receptor-mediated endocytosis and recycling in cells from mice with Gpr107 locus disruption.
Journal of cell science (2014-05-23)
Guo Ling Zhou, Soon-Young Na, Rasma Niedra, Brian Seed
ABSTRACT
GPR107 is a type III integral membrane protein that was initially predicted to be a member of the family of G-protein-coupled receptors. This report shows that deletion of Gpr107 leads to an embryonic lethal phenotype that is characterized by a reduction in cubilin transcript abundance and a decrease in the representation of multiple genes implicated in the cubilin-megalin endocytic receptor complex (megalin is also known as LRP2). Gpr107-null fibroblast cells exhibit reduced transferrin internalization, decreased uptake of low-density lipoprotein (LDL) receptor-related protein-1 (LRP1) cargo and resistance to toxins. Colocalization studies and proteomic analyses suggest that GPR107 associates with clathrin and the retromer protein VPS35 and that GPR107 might be responsible for the return of receptors to the plasma membrane from endocytic compartments. The highly selective deficits observed in Gpr107-null cells indicate that GPR107 interacts directly or indirectly with a limited subset of surface receptors.
Sigma-Aldrich
Anti-EEA1 antibody, Mouse monoclonal, clone EEA1-N19, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-LRP1 antibody produced in mouse, ~1.0 mg/mL, clone LRP1-11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)