MilliporeSigma
  • GPS2/KDM4A pioneering activity regulates promoter-specific recruitment of PPARγ.
GPS2/KDM4A pioneering activity regulates promoter-specific recruitment of PPARγ.
Cell reports (2014-06-24)
M Dafne Cardamone, Bogdan Tanasa, Michelle Chan, Carly T Cederquist, Jaclyn Andricovich, Michael G Rosenfeld, Valentina Perissi
ABSTRACT
Timely and selective recruitment of transcription factors to their appropriate DNA-binding sites represents a critical step in regulating gene activation; however, the regulatory strategies underlying each factor's effective recruitment to specific promoter and/or enhancer regions are not fully understood. Here, we identify an unexpected regulatory mechanism by which promoter-specific binding, and therefore function, of peroxisome proliferator-activator receptor γ (PPARγ) in adipocytes requires G protein suppressor 2 (GPS2) to prime the local chromatin environment via inhibition of the ubiquitin ligase RNF8 and stabilization of the H3K9 histone demethylase KDM4A/JMJD2. Integration of genome-wide profiling data indicates that the pioneering activity of GPS2/KDM4A is required for PPARγ-mediated regulation of a specific transcriptional program, including the lipolytic enzymes adipose triglyceride lipase (ATGL) and hormone-sensitive lipase (HSL). Hence, our findings reveal that GPS2 exerts a biologically important function in adipose tissue lipid mobilization by directly regulating ubiquitin signaling and indirectly modulating chromatin remodeling to prime selected genes for activation.
Sigma-Aldrich
Anti-RNF8 antibody produced in rabbit, IgG fraction of antiserum