MilliporeSigma
  • Changes of the major sperm maturation-associated epididymal protein HE5 (CD52) on human ejaculated spermatozoa during incubation.
Changes of the major sperm maturation-associated epididymal protein HE5 (CD52) on human ejaculated spermatozoa during incubation.
Molecular human reproduction (2001-06-23)
C H Yeung, F Pérez-Sánchez, S Schröter, C Kirchhoff, T G Cooper
ABSTRACT
HE5 (CD52) is a glycoprotein which is secreted by the epididymis and which becomes inserted onto maturing spermatozoa. We have previously shown that, in cynomolgus monkey spermatozoa, changes occur upon maturation rendering cryptic the epitope to the monoclonal antibody CAMPATH-1G; the recognition site is then re-exposed during incubation under capacitation conditions. The present study investigated human ejaculated spermatozoa during incubation under similar conditions, using monoclonal antibodies that recognize different epitopes of the HE5 molecule comprising parts of the N-glycan (2E5) or peptide segments, including (CAMPATH-1G) or excluding (097) the glycosylphosphatidylinositol (GPI) anchor, to reveal modifications of sperm surface HE5. Flow cytometric analysis showed equally high percentages (approximately 90%) of viable spermatozoa cross-reacting with the antibodies before and after 6 h incubation. However, during incubation, the staining intensity increased 57% with CAMPATH-1G, 31% with 097, but remained unchanged with 2E5. The lymphocyte CD52 antibody CF1D12 stained only approximately 10% of spermatozoa either before or after incubation. Western blotting of sperm protein extracts using lectins indicated an increase in the exposure of sialic acid residues of HE5 after incubation. These results suggest that during incubation in capacitating conditions, there is an opening up of the HE5 glycoprotein molecule, increasing accessibility of some sialic acid residues and of the core peptide, particularly the GPI anchor.