MilliporeSigma
  • VGLL4 targets a TCF4-TEAD4 complex to coregulate Wnt and Hippo signalling in colorectal cancer.
VGLL4 targets a TCF4-TEAD4 complex to coregulate Wnt and Hippo signalling in colorectal cancer.
Nature communications (2017-01-05)
Shi Jiao, Chuanchuan Li, Qian Hao, Haofei Miao, Lei Zhang, Lin Li, Zhaocai Zhou
ABSTRACT
Concerted co-regulation of multiple signalling pathways is crucial for tissue homoeostasis and tumorigenesis. Here we report that VGLL4, a previously identified YAP antagonist, also functions as a regulator of Wnt/β-catenin signalling. The expression of VGLL4 is significantly downregulated in clinical colorectal carcinoma (CRC) specimens, positively associated with patient survival rate, and inversely correlated with the expression of Wnt target genes in CRCs. Knockdown of VGLL4 enhances proliferation and tumour formation of CRC cells. A designed peptide mimicking the function of VGLL4 effectively inhibits CRC progression in a de novo mouse model. Mechanistically, TEAD4 associates with TCF4 to form a complex and cobind target genes. VGLL4 targets this TEAD4-TCF4 complex to interfere the functional interplay between TEAD4 and TCF4, suppressing the transactivation of TCF4. Collectively, our study indicates that Wnt/β-catenin and Hippo-YAP signalling are directly linked at transcription factor-level, and VGLL4 can target a TEAD4-TCF4 complex to co-regulate both pathways.
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human TEAD1
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture