MilliporeSigma
  • Spatial organization of embryonic stem cell responsiveness to autocrine gp130 ligands reveals an autoregulatory stem cell niche.
Spatial organization of embryonic stem cell responsiveness to autocrine gp130 ligands reveals an autoregulatory stem cell niche.
Stem cells (Dayton, Ohio) (2006-07-11)
Ryan E Davey, Peter W Zandstra
ABSTRACT
Highly ordered aggregates of cells, or niches, regulate stem cell fate. Specific tissue location need not be an obligatory requirement for a stem cell niche, particularly during embryogenesis, where cells exist in a dynamic environment. We investigated autoregulatory fixed-location-independent processes controlling cell fate by analyzing the spatial organization of embryonic stem cells (ESCs) using quantitative single-cell immunocytochemistry and a computational approach involving Delaunay triangulation. ESC colonies demonstrated radial organization of phosphorylated signal transducer and activator of transcription 3, Nanog, and Oct4 (among others) in the presence and absence of exogenous leukemia inhibitory factor (LIF). Endogenous self-renewal signaling resulted from autocrine non-LIF gp130 ligands, which buffered cells against differentiation upon exogenous LIF deprivation. Together with a radial organization of differential responsiveness to gp130 ligands within colonies, autocrine signaling produced a radial organization of self-renewal, generating a fixed-location-independent autoregulatory niche. These findings reveal fundamental properties of niches and elucidate mechanisms colonies of cells use to transition between fates during morphogenesis.
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody