MilliporeSigma
  • Reduced oxidative stress promotes NF-kappaB-mediated neuroprotective gene expression after transient focal cerebral ischemia: lymphocytotrophic cytokines and antiapoptotic factors.
Reduced oxidative stress promotes NF-kappaB-mediated neuroprotective gene expression after transient focal cerebral ischemia: lymphocytotrophic cytokines and antiapoptotic factors.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2006-07-27)
Yun Seon Song, Yong-Sun Lee, Purnima Narasimhan, Pak H Chan
ABSTRACT
Nuclear factor-kappa B (NF-kappaB) is activated by oxidative stress such as that induced by transient focal cerebral ischemia (tFCI). Whether NF-kappaB has a role in cell survival or death in stroke is a matter of debate. We proposed that the status of oxidative stress may determine its role in cell death or survival after focal ischemia. To characterize the coordinated expression of genes in NF-kappaB signaling after mild cerebral ischemia, we investigated the temporal profile of a NF-kappaB-pathway-focused DNA array after 30 mins of tFCI in wild-type (WT) mice and human copper/zinc-superoxide dismutase transgenic (SOD1 Tg) mice that had a significantly reduced level of superoxide. Differentially expressed genes among 96 NF-kappaB-related genes were further confirmed and compared in the WT and SOD1 Tg mice using quantitative polymerase chain reaction, Western blotting, and immunohistochemistry. Persistent upregulation of NF-kappaB seen at 7 days in the WT mice was decreased in the SOD1 Tg mice. Lymphocytotrophic cytokine genes such as interleukin-2, interleukin-12, and interferon-alpha1 were increased in the SOD1 Tg mice compared with the WT mice after tFCI. In addition, antiapoptosis factors bcl-2 and tumor necrosis factor receptor-associated factor 1 rapidly increased in the SOD1 Tg mice compared with the WT mice. This study indicates that reduced oxidative stress by SOD1 overexpression increased NF-kappaB-related rapid defenses, such as immune response and antiapoptosis factors, and prevented brain damage after tFCI-induced oxidative stress.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit