MilliporeSigma
  • Elevated body weight gain during the juvenile period alters neuropeptide Y-gonadotropin-releasing hormone circuitry in prepubertal heifers.
Elevated body weight gain during the juvenile period alters neuropeptide Y-gonadotropin-releasing hormone circuitry in prepubertal heifers.
Biology of reproduction (2014-12-17)
Bruna R C Alves, Rodolfo C Cardoso, Ligia D Prezotto, Jennifer F Thorson, Michelle Bedenbaugh, Sarah M Sharpton, Alain Caraty, Duane H Keisler, Luis O Tedeschi, Gary L Williams, Marcel Amstalden
ABSTRACT
Increased body weight (BW) gain during the juvenile period leads to early maturation of the reproductive neuroendocrine system. We investigated whether a nutritional regimen that advances the onset of puberty leads to alterations in the hypothalamic neuropeptide Y (NPY) circuitry that are permissive for enhanced gonadotropin-releasing hormone (GnRH) secretion. It was hypothesized that NPY mRNA and NPY projections to GnRH and kisspeptin neurons are reduced in heifers that gain BW at an accelerated rate, compared with a lower one, during the juvenile period. Heifers were weaned at approximately 4 mo of age and fed diets to promote relatively low (0.5 kg/day; low gain [LG]) or high (1.0 kg/day; high gain [HG]) rates of BW gain until 8.5 mo of age. Heifers that gained BW at a higher rate exhibited greater circulating concentrations of leptin and reduced overall NPY expression in the arcuate nucleus. The proportion of GnRH neurons in close apposition to NPY fibers and the magnitude of NPY projections to GnRH neurons located in the mediobasal hypothalamus were reduced in HG heifers. However, no differences in NPY projections to kisspeptin neurons in the arcuate nucleus were detected between HG and LG heifers. Results indicate that a reduction in NPY innervation of GnRH neurons, particularly at the level of the mediobasal hypothalamus, occurs in response to elevated BW gain during the juvenile period. This functional plasticity may facilitate early onset of puberty in heifers.
Sigma-Aldrich
Anti-Neuropeptide Y (NPY) antibody produced in rabbit, whole antiserum