MilliporeSigma
  • Transient association of MCM complex proteins with the nuclear matrix during initiation of mammalian DNA replication.
Transient association of MCM complex proteins with the nuclear matrix during initiation of mammalian DNA replication.
Cell cycle (Georgetown, Tex.) (2015-02-07)
Emma L Hesketh, John R P Knight, Rosemary H C Wilson, James P J Chong, Dawn Coverley
ABSTRACT
The minichromosome maintenance complex (MCM2-7) is the putative DNA helicase in eukaryotes, and essential for DNA replication. By applying serial extractions to mammalian cells synchronized by release from quiescence, we reveal dynamic changes to the sub-nuclear compartmentalization of MCM2 as cells pass through late G1 and early S phase, identifying a brief window when MCM2 becomes transiently attached to the nuclear-matrix. The data distinguish 3 states that correspond to loose association with chromatin prior to DNA replication, transient highly stable binding to the nuclear-matrix coincident with initiation, and a post-initiation phase when MCM2 remains tightly associated with chromatin but not the nuclear-matrix. The data suggests that functional MCM complex loading takes place at the nuclear-matrix.
Sigma-Aldrich
Monoclonal Anti-Cyclin A antibody produced in mouse, clone CY-A1, ascites fluid