MilliporeSigma
  • Targeting mammalian organelles with internalizing phage (iPhage) libraries.
Targeting mammalian organelles with internalizing phage (iPhage) libraries.
Nature protocols (2013-09-14)
Roberto Rangel, Andrey S Dobroff, Liliana Guzman-Rojas, Carolina C Salmeron, Juri G Gelovani, Richard L Sidman, Renata Pasqualini, Wadih Arap
ABSTRACT
Techniques that are largely used for protein interaction studies and the discovery of intracellular receptors, such as affinity-capture complex purification and the yeast two-hybrid system, may produce inaccurate data sets owing to protein insolubility, transient or weak protein interactions or irrelevant intracellular context. A versatile tool for overcoming these limitations, as well as for potentially creating vaccines and engineering peptides and antibodies as targeted diagnostic and therapeutic agents, is the phage-display technique. We have recently developed a new technology for screening internalizing phage (iPhage) vectors and libraries using a ligand/receptor-independent mechanism to penetrate eukaryotic cells. iPhage particles provide a unique discovery platform for combinatorial intracellular targeting of organelle ligands along with their corresponding receptors and for fingerprinting functional protein domains in living cells. Here we explain the design, cloning, construction and production of iPhage-based vectors and libraries, along with basic ligand-receptor identification and validation methodologies for organelle receptors. An iPhage library screening can be performed in ∼8 weeks.
Sigma-Aldrich
Anti-fd Bacteriophage antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Roche
Cell Proliferation Kit I (MTT)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%