MilliporeSigma
  • Glucocerebrosidase depletion enhances cell-to-cell transmission of α-synuclein.
Glucocerebrosidase depletion enhances cell-to-cell transmission of α-synuclein.
Nature communications (2014-08-27)
Eun-Jin Bae, Na-Young Yang, Miyoung Song, Cheol Soon Lee, Jun Sung Lee, Byung Chul Jung, He-Jin Lee, Seokjoong Kim, Eliezer Masliah, Sergio Pablo Sardi, Seung-Jae Lee
ABSTRACT
Deposition of α-synuclein aggregates occurs widely in the central and peripheral nervous systems in Parkinson's disease (PD). Although recent evidence has suggested that cell-to-cell transmission of α-synuclein aggregates is associated with the progression of PD, the mechanism by which α-synuclein aggregates spread remains undefined. Here, we show that α-synuclein aggregates are transmitted from cell to cell through a cycle involving uptake of external aggregates, co-aggregation with endogenous α-synuclein and exocytosis of the co-aggregates. Moreover, we find that glucocerebrosidase depletion, which has previously been strongly associated with PD and increased cognitive impairment, promotes propagation of α-synuclein aggregates. These studies define how α-synuclein aggregates spread among neuronal cells and may provide an explanation for how glucocerebrosidase mutations increase the risk of developing PD and other synucleinopathies.
Sigma-Aldrich
Anti-Glucocerebrosidase (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution