MilliporeSigma
  • Synapsin II desynchronizes neurotransmitter release at inhibitory synapses by interacting with presynaptic calcium channels.
Synapsin II desynchronizes neurotransmitter release at inhibitory synapses by interacting with presynaptic calcium channels.
Nature communications (2013-02-28)
Lucian Medrihan, Fabrizia Cesca, Andrea Raimondi, Gabriele Lignani, Pietro Baldelli, Fabio Benfenati
ABSTRACT
In the central nervous system, most synapses show a fast mode of neurotransmitter release known as synchronous release followed by a phase of asynchronous release, which extends over tens of milliseconds to seconds. Synapsin II (SYN2) is a member of the multigene synapsin family (SYN1/2/3) of synaptic vesicle phosphoproteins that modulate synaptic transmission and plasticity, and are mutated in epileptic patients. Here we report that inhibitory synapses of the dentate gyrus of Syn II knockout mice display an upregulation of synchronous neurotransmitter release and a concomitant loss of delayed asynchronous release. Syn II promotes γ-aminobutyric acid asynchronous release in a Ca(2+)-dependent manner by a functional interaction with presynaptic Ca(2+) channels, revealing a new role in synaptic transmission for synapsins.
Sigma-Aldrich
Anti-Calcium Channel (α1A Subunit) (P/Q-type of Voltage-gated Ca2+ Channel) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Calcium Channel (α1B Subunit) (N-type of Voltage-gated Ca2+ Channel) antibody produced in rabbit, affinity isolated antibody, lyophilized powder