MilliporeSigma
  • Chronic Acrylamide Exposure in Male Mice Results in Elevated DNA Damage in the Germline and Heritable Induction of CYP2E1 in the Testes.
Chronic Acrylamide Exposure in Male Mice Results in Elevated DNA Damage in the Germline and Heritable Induction of CYP2E1 in the Testes.
Biology of reproduction (2016-09-02)
Aimee L Katen, Caitlin G Chambers, Brett Nixon, Shaun D Roman
ABSTRACT
Acute acrylamide exposure in male rodents results in reduced reproductive performance and dominant lethality. However, the reproductive effects of low dose chronic exposure, which better reflects the nature of human exposure, remain far less certain. Human dietary consumption of acrylamide has been estimated at an average of 1-4 µg/kg bw/day. In order to simulate this exposure, male mice were provided with acrylamide (1 µg/ml) via their drinking water continuously for six months, which was equivalent to a human dose of 10.5 µg/ kg bw/day. This exposure regime increased DNA damage in the spermatozoa, without affecting a concomitant reduction in overall fertility. The offspring of acrylamide treated mice did not have an increased incidence of skin papilloma formation following the two-stage tumor induction protocol. However, the male offspring of acrylamide treated fathers had significantly increased levels of DNA damage in their spermatozoa, despite having had no direct toxicant exposure. It was also found that the F0, and most crucially, F1 mice had increased levels of CYP2E1 protein in their germ cells. This is significant as CYP2E1 is the sole enzyme responsible for conversion of acrylamide to its harmful metabolite glycidamide. This altered expression may be due to epigenetic alterations. Additionally, the F0 and F1 mice had increased oxidative adducts in the DNA of their germ cells, which was hypothesized to arise as a byproduct of increased CYP2E1 activity. Therefore, chronic paternal acrylamide exposure in mice has consequences for their offspring, and raises concerns for the effects of acrylamide exposure in the human population and the accumulated effects with multiple generations of exposure.
Sigma-Aldrich
Goat Anti-Rabbit IgG Peroxidase Conjugate, lyophilized, Calbiochem®
Sigma-Aldrich
Anti-8-Hydroxydeoxyguanosine Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution