MilliporeSigma
NRF2 regulates serine biosynthesis in non-small cell lung cancer.
Nature genetics (2015-10-21)
Gina M DeNicola, Pei-Hsuan Chen, Edouard Mullarky, Jessica A Sudderth, Zeping Hu, David Wu, Hao Tang, Yang Xie, John M Asara, Kenneth E Huffman, Ignacio I Wistuba, John D Minna, Ralph J DeBerardinis, Lewis C Cantley
ABSTRACT
Tumors have high energetic and anabolic needs for rapid cell growth and proliferation, and the serine biosynthetic pathway was recently identified as an important source of metabolic intermediates for these processes. We integrated metabolic tracing and transcriptional profiling of a large panel of non-small cell lung cancer (NSCLC) cell lines to characterize the activity and regulation of the serine/glycine biosynthetic pathway in NSCLC. Here we show that the activity of this pathway is highly heterogeneous and is regulated by NRF2, a transcription factor frequently deregulated in NSCLC. We found that NRF2 controls the expression of the key serine/glycine biosynthesis enzyme genes PHGDH, PSAT1 and SHMT2 via ATF4 to support glutathione and nucleotide production. Moreover, we show that expression of these genes confers poor prognosis in human NSCLC. Thus, a substantial fraction of human NSCLCs activates an NRF2-dependent transcriptional program that regulates serine and glycine metabolism and is linked to clinical aggressiveness.
Sigma-Aldrich
4-Ethylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
Anti-PHGDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-NQO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit