MilliporeSigma
  • Death-associated protein kinase 1 is an IRF3/7-interacting protein that is involved in the cellular antiviral immune response.
Death-associated protein kinase 1 is an IRF3/7-interacting protein that is involved in the cellular antiviral immune response.
Cellular & molecular immunology (2014-02-18)
Jing Zhang, Ming-Ming Hu, Hong-Bing Shu, Shu Li
ABSTRACT
Interferon regulatory factor (IRF) 7 has been demonstrated to be a master regulator of virus-induced type I interferon production (IFN), and it plays a central role in the innate immune response against viruses. Here, we identified death-associated protein kinase 1 (DAPK1) as an IRF7-interacting protein by tandem affinity purification (TAP). Viral infection induced DAPK1-IRF7 and DAPK1-IRF3 interactions and overexpression of DAPK1 enhanced virus-induced activation of the interferon-stimulated response element (ISRE) and IFN-β promoters and the expression of the IFNB1 gene. Knockdown of DAPK1 attenuated the induction of IFNB1 and RIG-I expression triggered by viral infection or IFN-β, and they were enhanced by viral replication. In addition, viral infection or IFN-β treatment induced the expression of DAPK1. IFN-β treatment also activated DAPK1 by decreasing its phosphorylation level at serine 308. Interestingly, the involvement of DAPK1 in virus-induced signaling was independent of its kinase activity. Therefore, our study identified DAPK1 as an important regulator of the cellular antiviral response.
Sigma-Aldrich
Anti-DAPK1 antibody produced in rabbit, affinity isolated antibody