MilliporeSigma
  • Regional and age-related differences in GAD67 expression of parvalbumin- and calbindin-expressing neurons in the rhesus macaque auditory midbrain and brainstem.
Regional and age-related differences in GAD67 expression of parvalbumin- and calbindin-expressing neurons in the rhesus macaque auditory midbrain and brainstem.
The Journal of comparative neurology (2014-08-06)
D T Gray, J R Engle, M L Rudolph, G H Recanzone
ABSTRACT
Neurons expressing the calcium binding proteins (CaBPs) parvalbumin (PV) and calbindin (CB) have shown age-related density changes throughout the ascending auditory system of both rodents and macaque monkeys. In the cerebral cortex, neurons expressing these CaBPs express markers of γ-aminobutyric acidergic neurotransmission, such as GAD67, and have well-understood physiological response properties. Recent evidence suggests that, in the rodent auditory brainstem, CaBP-containing cells do not express GAD67. It is unknown whether PV- and CB-containing cells in subcortical auditory structures of macaques similarly do not express GAD67, and a better understanding of the neurotransmission of neurons expressing these proteins is necessary for understanding the age-related changes in their density throughout the macaque auditory system. This was investigated with immunofluorescent double-labeling techniques to coregister PV- and CB-expressing neurons with GAD67 in the superior olivary complex and the inferior colliculus of young and aged rhesus macaques. The proportions of GAD67-expressing PV- and CB-positive neurons were computed with unbiased sampling techniques. Our results indicate that between 42% and 62% of PV- and CB-positive neurons in the auditory brainstem and midbrain express GAD67, which is significantly less than in the cerebrum. In general, fewer PV(+) neurons and more CB(+) neurons expressed GAD67 as a function of age. These results demonstrate that the inhibitory molecular profile of PV- and CB-expressing neurons can change with age in subcortical auditory structures and that these neurons are distinct from the well-described inhibitory interneurons that express these proteins in the cerebral cortex.
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid
Sigma-Aldrich
Anti-GAD1 antibody produced in rabbit, affinity isolated antibody