MilliporeSigma
  • Conserved expression of the GPR151 receptor in habenular axonal projections of vertebrates.
Conserved expression of the GPR151 receptor in habenular axonal projections of vertebrates.
The Journal of comparative neurology (2014-08-15)
Jonas Broms, Beatriz Antolin-Fontes, Anders Tingström, Ines Ibañez-Tallon
ABSTRACT
The habenula is a phylogenetically conserved brain structure in the epithalamus. It is a major node in the information flow between fronto-limbic brain regions and monoaminergic brainstem nuclei, and is thus anatomically and functionally ideally positioned to regulate emotional, motivational, and cognitive behaviors. Consequently, the habenula may be critically important in the pathophysiology of psychiatric disorders such as addiction and depression. Here we investigated the expression pattern of GPR151, a G protein-coupled receptor (GPCR), whose mRNA has been identified as highly and specifically enriched in habenular neurons by in situ hybridization and translating ribosome affinity purification (TRAP). In the present immunohistochemical study we demonstrate a pronounced and highly specific expression of the GPR151 protein in the medial and lateral habenula of rodent brain. Specific expression was also seen in efferent habenular fibers projecting to the interpeduncular nucleus, the rostromedial tegmental area, the rhabdoid nucleus, the mesencephalic raphe nuclei, and the dorsal tegmental nucleus. Using confocal microscopy and quantitative colocalization analysis, we found that GPR151-expressing axons and terminals overlap with cholinergic, substance P-ergic, and glutamatergic markers. Virtually identical expression patterns were observed in rat, mouse, and zebrafish brains. Our data demonstrate that GPR151 is highly conserved, specific for a subdivision of the habenular neurocircuitry, and constitutes a promising novel target for psychiatric drug development.
Sigma-Aldrich
Anti-GPR151 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-Tyrosine Hydroxylase antibody produced in mouse, clone TH-2, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Tryptophan Hydroxylase antibody produced in mouse, clone WH-3, ascites fluid
Sigma-Aldrich
Anti-Tryptophan Hydroxylase Antibody, Chemicon®, from sheep
Sigma-Aldrich
Anti-GPR151 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat