MilliporeSigma
  • Golgi self-correction generates bioequivalent glycans to preserve cellular homeostasis.
Golgi self-correction generates bioequivalent glycans to preserve cellular homeostasis.
eLife (2016-06-09)
Haik Mkhikian, Christie-Lynn Mortales, Raymond W Zhou, Khachik Khachikyan, Gang Wu, Stuart M Haslam, Patil Kavarian, Anne Dell, Michael Demetriou
ABSTRACT
Essential biological systems employ self-correcting mechanisms to maintain cellular homeostasis. Mammalian cell function is dynamically regulated by the interaction of cell surface galectins with branched N-glycans. Here we report that N-glycan branching deficiency triggers the Golgi to generate bioequivalent N-glycans that preserve galectin-glycoprotein interactions and cellular homeostasis. Galectins bind N-acetyllactosamine (LacNAc) units within N-glycans initiated from UDP-GlcNAc by the medial-Golgi branching enzymes as well as the trans-Golgi poly-LacNAc extension enzyme β1,3-N-acetylglucosaminyltransferase (B3GNT). Marginally reducing LacNAc content by limiting N-glycans to three branches results in T-cell hyperactivity and autoimmunity; yet further restricting branching does not produce a more hyperactive state. Rather, new poly-LacNAc extension by B3GNT maintains galectin binding and immune homeostasis. Poly-LacNAc extension is triggered by redistribution of unused UDP-GlcNAc from the medial to trans-Golgi via inter-cisternal tubules. These data demonstrate the functional equivalency of structurally dissimilar N-glycans and suggest a self-correcting feature of the Golgi that sustains cellular homeostasis.
Sigma-Aldrich
Anti-TGN46 antibody, Mouse monoclonal, clone TGN46-52, purified from hybridoma cell culture