MilliporeSigma
  • Covalent targeting of remote cysteine residues to develop CDK12 and CDK13 inhibitors.
Covalent targeting of remote cysteine residues to develop CDK12 and CDK13 inhibitors.
Nature chemical biology (2016-08-30)
Tinghu Zhang, Nicholas Kwiatkowski, Calla M Olson, Sarah E Dixon-Clarke, Brian J Abraham, Ann K Greifenberg, Scott B Ficarro, Jonathan M Elkins, Yanke Liang, Nancy M Hannett, Theresa Manz, Mingfeng Hao, Bartlomiej Bartkowiak, Arno L Greenleaf, Jarrod A Marto, Matthias Geyer, Alex N Bullock, Richard A Young, Nathanael S Gray
ABSTRACT
Cyclin-dependent kinases 12 and 13 (CDK12 and CDK13) play critical roles in the regulation of gene transcription. However, the absence of CDK12 and CDK13 inhibitors has hindered the ability to investigate the consequences of their inhibition in healthy cells and cancer cells. Here we describe the rational design of a first-in-class CDK12 and CDK13 covalent inhibitor, THZ531. Co-crystallization of THZ531 with CDK12-cyclin K indicates that THZ531 irreversibly targets a cysteine located outside the kinase domain. THZ531 causes a loss of gene expression with concurrent loss of elongating and hyperphosphorylated RNA polymerase II. In particular, THZ531 substantially decreases the expression of DNA damage response genes and key super-enhancer-associated transcription factor genes. Coincident with transcriptional perturbation, THZ531 dramatically induced apoptotic cell death. Small molecules capable of specifically targeting CDK12 and CDK13 may thus help identify cancer subtypes that are particularly dependent on their kinase activities.
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2) Antibody, clone 3E10, clone 3E10, from rat
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12, clone 4E12, from rat
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-5) Antibody, clone 3E8, clone 3E8, from rat
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid