MilliporeSigma
  • Shear stress induces synthetic-to-contractile phenotypic modulation in smooth muscle cells via peroxisome proliferator-activated receptor alpha/delta activations by prostacyclin released by sheared endothelial cells.
Shear stress induces synthetic-to-contractile phenotypic modulation in smooth muscle cells via peroxisome proliferator-activated receptor alpha/delta activations by prostacyclin released by sheared endothelial cells.
Circulation research (2009-07-25)
Min-Chien Tsai, Lihong Chen, Jing Zhou, Zhihui Tang, Tzu-Fang Hsu, Ying Wang, Yu-Tsung Shih, Hsin-Hsin Peng, Nanping Wang, Youfei Guan, Shu Chien, Jeng-Jiann Chiu
ABSTRACT
Phenotypic modulation of smooth muscle cells (SMCs), which are located in close proximity to endothelial cells (ECs), is critical in regulating vascular function. The role of flow-induced shear stress in the modulation of SMC phenotype has not been well defined. The objective was to elucidate the role of shear stress on ECs in modulating SMC phenotype and its underlying mechanism. Application of shear stress (12 dyn/cm2) to ECs cocultured with SMCs modulated SMC phenotype from synthetic to contractile state, with upregulation of contractile markers, downregulation of proinflammatory genes, and decreased percentage of cells in the synthetic phase. Treating SMCs with media from sheared ECs induced peroxisome proliferator-activated receptor (PPAR)-alpha, -delta, and -gamma ligand binding activities; transfecting SMCs with specific small interfering (si)RNAs of PPAR-alpha and -delta, but not -gamma, inhibited shear induction of contractile markers. ECs exposed to shear stress released prostacyclin (PGI2). Transfecting ECs with PGI2 synthase-specific siRNA inhibited shear-induced activation of PPAR-alpha/delta, upregulation of contractile markers, downregulation of proinflammatory genes, and decrease in percentage of SMCs in synthetic phase. Mice with PPAR-alpha deficiency (compared with control littermates) showed altered SMC phenotype toward a synthetic state, with increased arterial contractility in response to angiotensin II. These results indicate that laminar shear stress induces synthetic-to-contractile phenotypic modulation in SMCs through the activation of PPAR-alpha/delta by the EC-released PGI2. Our findings provide insights into the mechanisms underlying the EC-SMC interplays and the protective homeostatic function of laminar shear stress in modulating SMC phenotype.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Caldesmon (Smooth) antibody produced in mouse, clone hHCD, ascites fluid