MilliporeSigma
  • Definitive identification of human T cells in formalin fixed paraffin wax embedded tissue.
Definitive identification of human T cells in formalin fixed paraffin wax embedded tissue.
Journal of clinical pathology (1988-08-01)
N D Francis, D M Clark, A W Boylston
ABSTRACT
The use of a new monoclonal antibody, BF1, which reacts in routinely fixed tissues, directed against the beta chain of the T cell antigen receptor was assessed to determine its reactivity in a variety of normal tissues as well as in a number of B and T cell tumours and other lymphoid disorders. It reacted with all five of the unequivocal T cell tumours tested. BF1 will have widespread use in the assessment of lymphoid tissues in patients with the acquired immune deficiency syndrome (AIDS) and the differential diagnosis of tumours.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) antibody produced in goat, affinity isolated antibody, lyophilized powder