MilliporeSigma
  • The anti-tumor effects of calorie restriction are correlated with reduced oxidative stress in ENU-induced gliomas.
The anti-tumor effects of calorie restriction are correlated with reduced oxidative stress in ENU-induced gliomas.
Pathobiology of aging & age related diseases (2011-01-01)
Megan A Mahlke, Lisa A Cortez, Melanie A Ortiz, Marisela Rodriguez, Koji Uchida, Mark K Shigenaga, Shuko Lee, Yiquang Zhang, Kaoru Tominaga, Gene B Hubbard, Yuji Ikeno
ABSTRACT
The anti-tumor effects of calorie restriction (CR) and the possible underlying mechanisms were investigated using ethylnitrosourea (ENU)-induced glioma in rats. ENU was given transplacentally at gestational day 15, and male offspring were used in this experiment. The brain from 4-, 6-, and 8-month-old rats fed either ad libitum (AL) or calorie-restricted diets (40% restriction of total calories compared to AL rats) was studied. Tumor burden was assessed by comparing the number and size of gliomas present in sections of the brain. Immunohistochemical analysis was used to document lipid peroxidation [4-hydroxy-2-nonenal (HNE) and malondialdehyde (MDA)], protein oxidation (nitrotyrosine), glycation and AGE formation [methylglyoxal (MG) and carboxymethyllysine (CML)], cell proliferation activity [proliferating cell nuclear antigen (PCNA)], cell death [single-stranded DNA (ssDNA)], presence of thioredoxin 1 (Trx1), and presence of heme oxygenase-1 (HO-1) associated with the development of gliomas. The results showed that the number of gliomas did not change with age in the AL groups; however, the average size of the gliomas was significantly larger in the 8-month-old group compared to that of the younger groups. Immunopositivity was observed mainly in tumor cells and reactive astrocytes in all histological types of ENU-induced glioma. Immunopositive areas for HNE, MDA, nitrotyrosine, MG, CML, HO-1, and Trx1 increased with the growth of gliomas. The CR group showed both reduced number and size of gliomas, and tumors exhibited less accumulation of oxidative damage, decreased formation of glycated end products, and a decreased presence of HO-1 and Trx1 compared to the AL group. Furthermore, gliomas of the CR group showed less PCNA positive and more ssDNA positive cells, which are correlated to the retarded growth of tumors. Interestingly, we also discovered that the anti-tumor effects of CR were associated with decreased hypoxia-inducible factor-1α (HIF-1α) levels in normal brain tissue. Our results are very exciting because they not only demonstrate the anti-tumor effects of CR in gliomas, but also indicate the possible underlying mechanisms, i.e. anti-tumor effects of CR observed in this investigation are associated with reduced accumulation of oxidative damage, decreased formation of glycated end products, decreased presence of HO-1 and Trx1, reduced cell proliferation and increased apoptosis, and decreased levels of HIF-1α.
Sigma-Aldrich
Anti-Heme Oxygenase-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Thioredoxin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution