MilliporeSigma
  • Massive renal urothelial carcinoma with renal vein tumor thrombus, pancreatic infiltration and adrenal metastasis: A case report.
Massive renal urothelial carcinoma with renal vein tumor thrombus, pancreatic infiltration and adrenal metastasis: A case report.
Oncology letters (2016-07-23)
Tao Li, Liang Gao, Weilu Wu, Peng Chen, Siyuan Bu, Qiang Wei, Lu Yang
ABSTRACT
A 49-year-old female patient presented with a massive left renal tumor, recurrent left flank pain and gross hematuria. The tumor was accompanied by a renal vein tumor thrombus, pancreatic infiltration and a solitary adrenal metastasis. Radical nephrectomy, distal pancreatectomy, ipsilateral adrenalectomy and splenectomy were performed. Histopathological examination suggested high-grade urothelial carcinoma (UC); however, tumor recurrence and multiple metastases were detected only 3 months after the surgery, and the patient succumbed during follow-up 1 month later. To the best of our knowledge, this is the first case of renal UC of such advanced stage with renal vein tumor thrombus, pancreatic infiltration and a solitary adrenal metastasis.
Sigma-Aldrich
Monoclonal Anti-Cytokeratin, pan antibody produced in mouse, clone C-11, ascites fluid