MilliporeSigma
  • Carbenoxolone blocks endotoxin-induced protein kinase R (PKR) activation and high mobility group box 1 (HMGB1) release.
Carbenoxolone blocks endotoxin-induced protein kinase R (PKR) activation and high mobility group box 1 (HMGB1) release.
Molecular medicine (Cambridge, Mass.) (2013-07-10)
Wei Li, Jianhua Li, Andrew E Sama, Haichao Wang
ABSTRACT
The pathogen- and damage-associated molecular patterns (for example, bacterial endotoxin and adenosine 5'-triphosphate [ATP]) activate the double-stranded RNA-activated protein kinase R (PKR) to trigger the inflammasome-dependent high mobility group box 1 (HMGB1) release. Extracellular ATP contributes to the inflammasome activation through binding to the plasma membrane purinergic P2X7 receptor (P2X7R), triggering the opening of P2X7R channels and the pannexin-1 (panx-1) hemichannels permeable for larger molecules up to 900 daltons. It was previously unknown whether panx-1 channel blockers can abrogate lipopolysaccharide (LPS)-induced PKR activation and HMGB1 release in innate immune cells. Here we demonstrated that a major gancao (licorice) component (glycyrrhizin, or glycyrrhizic acid) derivative, carbenoxolone (CBX), dose dependently abrogated LPS-induced HMGB1 release in macrophage cultures with an estimated IC50 ≈ 5 μmol/L. In an animal model of polymicrobial sepsis (induced by cecal ligation and puncture [CLP]), repetitive CBX administration beginning 24 h after CLP led to a significant reduction of circulating and peritoneal HMGB1 levels, and promoted a significant increase in animal survival rates. As did P2X7R antagonists (for example, oxidized ATP, oATP), CBX also effectively attenuated LPS-induced P2X7R/panx-1 channel activation (as judged by Lucifer Yellow dye uptake) and PKR phosphorylation in primary peritoneal macrophages. Collectively, these results suggested that CBX blocks LPS-induced HMGB1 release possibly through impairing PKR activation, supporting the involvement of PKR in the regulation of HMGB1 release.
Sigma-Aldrich
Anti-phospho-PKR (Thr451) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture