MilliporeSigma
  • Human monoclonal anti-protective antigen antibody completely protects rabbits and is synergistic with ciprofloxacin in protecting mice and guinea pigs against inhalation anthrax.
Human monoclonal anti-protective antigen antibody completely protects rabbits and is synergistic with ciprofloxacin in protecting mice and guinea pigs against inhalation anthrax.
Infection and immunity (2006-01-24)
Johnny W Peterson, Jason E Comer, David M Noffsinger, Autumn Wenglikowski, Kristin G Walberg, Bagram M Chatuev, Ashok K Chopra, Lawrence R Stanberry, Angray S Kang, Wolfgang W Scholz, Jagadish Sircar
ABSTRACT
Prevention of inhalation anthrax requires early and extended antibiotic therapy, and therefore, alternative treatment strategies are needed. We investigated whether a human monoclonal antibody (AVP-21D9) to protective antigen (PA) would protect mice, guinea pigs, and rabbits against anthrax. Control animals challenged with Bacillus anthracis Ames spores by the intranasal route died within 3 to 7 days. AVP-21D9 alone provided minimal protection against anthrax in the murine model, but its efficacy was notably better in guinea pigs. When Swiss-Webster mice, challenged with five 50% lethal doses (LD50s) of anthrax spores, were given a single 16.7-mg/kg of body weight AVP-21D9 antibody dose combined with ciprofloxacin (30 mg/kg/day for 6 days) 24 h after challenge, 100% of the mice were protected for more than 30 days, while ciprofloxacin or AVP-21D9 alone showed minimal protection. Similarly, when AVP-21D9 antibody (10 to 50 mg/kg) was combined with a low, nonprotective dose of ciprofloxacin (3.7 mg/kg/day) and administered to guinea pigs for 6 days, synergistic protection against anthrax was observed. In contrast, a single dose of AVP-21D9 antibody (1, 5, 10, or 20 mg/kg) but not 0.2 mg/kg alone completely protected rabbits against challenge with 100 LD50s of B. anthracis Ames spores, and 100% of the rabbits survived rechallenge. Further, administration of AVP-21D9 (10 mg/kg) to rabbits at 0, 6, and 12 h after challenge with anthrax spores resulted in 100% survival; however, delay of antibody treatment by 24 and 48 h reduced survival to 80% and 60%, respectively. Serological analysis of sera from various surviving animals 30 days postprimary infection showed development of a species-specific PA enzyme-linked immunosorbent assay antibody titer that correlated with protection against reinfection. Taken together, the effectiveness of human anti-PA antibody alone or in combination with low ciprofloxacin levels may provide the basis for an improved strategy for prophylaxis or treatment following inhalation anthrax infection.
Sigma-Aldrich
IgG1, Kappa from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)