MilliporeSigma
  • Oxidative lipid modification of nicastrin enhances amyloidogenic γ-secretase activity in Alzheimer's disease.
Oxidative lipid modification of nicastrin enhances amyloidogenic γ-secretase activity in Alzheimer's disease.
Aging cell (2012-03-13)
A-Ryeong Gwon, Jong-Sung Park, Thiruma V Arumugam, Yong-Kook Kwon, Sic L Chan, Seol-Hee Kim, Sang-Ha Baik, Sunghee Yang, Young-Kwang Yun, Yuri Choi, Saerom Kim, Sung-Chun Tang, Dong-Hoon Hyun, Aiwu Cheng, Charles E Dann, Michel Bernier, Jaewon Lee, William R Markesbery, Mark P Mattson, Dong-Gyu Jo
ABSTRACT
The cause of elevated level of amyloid β-peptide (Aβ42) in common late-onset sporadic [Alzheimer's disease (AD)] has not been established. Here, we show that the membrane lipid peroxidation product 4-hydroxynonenal (HNE) is associated with amyloid and neurodegenerative pathologies in AD and that it enhances γ-secretase activity and Aβ42 production in neurons. The γ-secretase substrate receptor, nicastrin, was found to be modified by HNE in cultured neurons and in brain specimens from patients with AD, in which HNE-nicastrin levels were found to be correlated with increased γ-secretase activity and Aβ plaque burden. Furthermore, HNE modification of nicastrin enhanced its binding to the γ-secretase substrate, amyloid precursor protein (APP) C99. In addition, the stimulation of γ-secretase activity and Aβ42 production by HNE were blocked by an HNE-scavenging histidine analog in a 3xTgAD mouse model of AD. These findings suggest a specific molecular mechanism by which oxidative stress increases Aβ42 production in AD and identify HNE as a novel therapeutic target upstream of the γ-secretase cleavage of APP.
Sigma-Aldrich
Anti-Nicastrin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Amyloid Precursor Protein Antibody, universal, serum, Chemicon®