MilliporeSigma
  • Rational design of a protein that binds integrin αvβ3 outside the ligand binding site.
Rational design of a protein that binds integrin αvβ3 outside the ligand binding site.
Nature communications (2016-06-01)
Ravi Chakra Turaga, Lu Yin, Jenny J Yang, Hsiauwei Lee, Ivaylo Ivanov, Chunli Yan, Hua Yang, Hans E Grossniklaus, Siming Wang, Cheng Ma, Li Sun, Zhi-Ren Liu
ABSTRACT
Integrin αvβ3 expression is altered in various diseases and has been proposed as a drug target. Here we use a rational design approach to develop a therapeutic protein, which we call ProAgio, that binds to integrin αvβ3 outside the classical ligand-binding site. We show ProAgio induces apoptosis of integrin αvβ3-expressing cells by recruiting and activating caspase 8 to the cytoplasmic domain of integrin αvβ3. ProAgio also has anti-angiogenic activity and strongly inhibits growth of tumour xenografts, but does not affect the established vasculature. Toxicity analyses demonstrate that ProAgio is not toxic to mice. Our study reports a new integrin-targeting agent with a unique mechanism of action, and provides a template for the development of integrin-targeting therapeutics.
Sigma-Aldrich
Anti-Integrin beta-3 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid