MilliporeSigma
Wt1 and epicardial fate mapping.
Circulation research (2012-06-14)
Carsten Rudat, Andreas Kispert
ABSTRACT
The embryonic epicardium is a crucial cell source of the cardiac fibrous skeleton as well as of the coronary system. Genetic lineage tracing systems based on Wt1 regulatory sequences provided evidence that epicardium-derived cells also adopt a myocardial fate in the mouse. To define the adequacy of Wt1-based lineage tracing systems for epicardial fate mapping. Using in situ hybridization analysis and immunofluorescence on tissue sections, we detected endogenous expression of Wt1 mRNA and Wt1 protein in the proepicardium and epicardium and also in endothelial cells throughout cardiogenesis. Expression analysis of a sensitive GFP reporter showed that recombination mediated by cre recombinase in the Wt1(creEGFP) line occurs randomly and sporadically in all cells of the embryo. Recombination in cardiomyocytes was found in the linear heart tube before establishment of a (pro)epicardium. In contrast, the tamoxifen-inducible Wt1(creERT2) mouse line mediated poor and variable recombination in the epicardium. Recombination in cardiomyocytes was not detected in this case. Frequently used Wt1 based cre-mediated lineage tracing systems are not suitable for epicardial fate mapping because of endogenous endothelial expression of Wt1, ectopic recombination (Wt1(creEGFP)), and poor recombination efficiency (Wt1(creERT2)) in the developing heart. We conclude that claims of a cardiomyocyte fate of epicardial cells in the mouse are not substantiated.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture