MilliporeSigma
  • Cdk5 and its substrates, Dcx and p27kip1, regulate cytoplasmic dilation formation and nuclear elongation in migrating neurons.
Cdk5 and its substrates, Dcx and p27kip1, regulate cytoplasmic dilation formation and nuclear elongation in migrating neurons.
Development (Cambridge, England) (2014-09-04)
Yoshiaki V Nishimura, Mima Shikanai, Mikio Hoshino, Toshio Ohshima, Yo-ichi Nabeshima, Ken-Ichi Mizutani, Koh-Ichi Nagata, Kazunori Nakajima, Takeshi Kawauchi
ABSTRACT
Neuronal migration is crucial for development of the mammalian-specific six-layered cerebral cortex. Migrating neurons are known to exhibit distinct features; they form a cytoplasmic dilation, a structure specific to migrating neurons, at the proximal region of the leading process, followed by nuclear elongation and forward movement. However, the molecular mechanisms of dilation formation and nuclear elongation remain unclear. Using ex vivo chemical inhibitor experiments, we show here that rottlerin, which is widely used as a specific inhibitor for PKCδ, suppresses the formation of a cytoplasmic dilation and nuclear elongation in cortical migrating neurons. Although our previous study showed that cortical neuronal migration depends on Jnk, another downstream target of rottlerin, Jnk inhibition disturbs only the nuclear elongation and forward movement, but not the dilation formation. We found that an unconventional cyclin-dependent kinase, Cdk5, is a novel downstream target of rottlerin, and that pharmacological or knockdown-mediated inhibition of Cdk5 suppresses both the dilation formation and nuclear elongation. We also show that Cdk5 inhibition perturbs endocytic trafficking as well as microtubule organization, both of which have been shown to be required for dilation formation. Furthermore, knockdown of Dcx, a Cdk5 substrate involved in microtubule organization and membrane trafficking, or p27(kip1), another Cdk5 substrate involved in actin and microtubule organization, disturbs the dilation formation and nuclear elongation. These data suggest that Cdk5 and its substrates, Dcx and p27(kip1), characterize migrating neuron-specific features, cytoplasmic dilation formation and nuclear elongation in the mouse cerebral cortex, possibly through the regulation of microtubule organization and an endocytic pathway.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit